Learn More
Greiner Bio-One CELLCOAT™ 96-well, Poly-D Lysine-Treated, Flat-Bottom Microplate
GREENER_CHOICE
Catalog No. 07000192 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
White, Clear Bottom
Catalog No. Color
07-000-192 White, Clear Bottom
1 options
Catalog No. 07-000-192 Supplier Greiner Bio-One Supplier No. 655944
Add to Cart
Add to Cart
Color:White, Clear Bottom; Sterility:Non-sterile; Lid:With Lid
  • Reduced-serum or serum-free cultivation
  • Cell differentiation and neuron growth
  • Promotion of cell adhesion, proliferation and growth of transfected cell lines
  • Increase in isolation and cultivation efficiency
  • Consist
TRUSTED_SUSTAINABILITY
Specifications
Lid With Lid
Array 8 x 12
Autoclavable Not Autoclavable
Packaging 5/Pk., 20/Cs.
Color White, Clear Bottom
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 25 to 340 μL
Sterility Non-sterile
Surface Treatment Poly-D-Lysine
Total Volume (Metric) Well 392 μL
Well Shape Flat Bottom (F-well)
Max. RCF 4800 x g (swinging bucket rotor)
No. per Case 20
No. per Pack 5
Quantity 20/Pk.
Temperature Range (Metric) Room Temperature
Well Design Chimney