Learn More
Greiner Bio-One CELLCOAT™ 96-well, Poly-D Lysine-Treated, Flat-Bottom Microplate
GREENER_CHOICE
Catalog No. 07000193 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
White
Catalog No. Color
07-000-193 White
1 options
Catalog No. 07-000-193 Supplier Greiner Bio-One Supplier No. 655945
Add to Cart
Add to Cart
Color:White; Sterility:Non-sterile; Lid:With Lid
Alphanumeric well coding
TRUSTED_SUSTAINABILITY
Specifications
Lid With Lid
Array 8 x 12
Color White
Material Polystyrene
No. of Wells 96
Sterility Non-sterile
Surface Treatment Poly-D-Lysine
Well Shape Flat Bottom (F-well)
Quantity 20/Cs.