Learn More
Description
- Coated with poly-D-lysine
- Improved adhesion and proliferation of primary cells, various cell lines
- Sterile
Specifications
Specifications
| Bottom Shape | Flat |
| Color | Clear |
| Sterility | Sterile |
| Surface Treatment | Poly-D-Lysine |
| Type | Multi-well Cell Culture Plate |
| Well Count | 12 |
| For Use With (Application) | Serum-free and serum-reduced cell cultivation and transfection |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.