Learn More
Description
Greiner Bio-One CELLCOAT Poly-D-lysine Coated Cell Culture Vessel is used for cultivation of transfected cells and fastidious cell lines, as well as serum-free or -reduced cell culture.
- Source: Synthetic
- Cultivation of neuronal cells, transfected cells and other fastidious cell lines
- Cultivation in serum-free and serumreduced medium
- Improvement of cell attachment
Specifications
Specifications
| Closure Color | Clear |
| Closure Included | Closure Included |
| Closure Material | Polystyrene |
| Bottom Shape | Flat |
| Capacity (Metric) | 28 mL |
| Color | Clear |
| Culture Area | 21 cm² |
| Material | Polystyrene |
| Shape | Round |
| Sterility | Non-sterile |
| Show More |
For research use only. Not for use in diagnostic procedures, unless otherwise indicated
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.