Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One CELLCOAT™ Poly-D-Lysine Protein Coated Cell Culture Multiwell Plates

Catalog No. p-7075426 Shop All Greiner Bio-One Products
Change view
Click to view available options
Culture Area:
1.9 sq. cm
9.6 sq. cm
Capacity (Metric):
3.3 mL
16.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Culture Area Capacity (Metric)
07-000-182 1.9 sq. cm 3.3 mL
07-000-185 9.6 sq. cm 16.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 07-000-182 Supplier Greiner Bio-One Supplier No. 662940
Only null left
Add to Cart
Add to Cart

Ideal for low-serum, serum-free cultivation and cell differentiation. Greiner Bio-One CELLCOAT™ Poly-D-Lysine Protein Coated Cell Culture Multiwell Plates helps in promotion of cell adhesion and proliferation.

Poly-D-lysine (PDL) are synthetic molecules which increase cell adhesion to vessels by altering the net charge of the surface without stimulating biological activity.
  • Coated with Poly-D-Lysine synthetic protein (75-150 kDa)
  • For promotion of cell adhesion, proliferation and growth of transfected cell lines, neuronal cell lines, primary neurons and glial cells
  • Ready-to-use products
  • Includes lid with condensation rings
TRUSTED_SUSTAINABILITY

Specifications

Closure Color Clear
Closure Included Closure Included
Closure Material Polystyrene
Autoclavable Not Autoclavable
Bottom Shape Flat
Capacity (Metric) 3.3 mL
Color Clear
Culture Area 1.9 sq. cm
Material Polystyrene
Shape Rectangular
Sterility Non-sterile
Surface Treatment Poly-L-Lysine
Type Multi-well Cell Culture Plate
Volume (Metric) Working 0.5 to 1.0 mL
Well Count 24
Includes Lid
No. per Case 50
No. per Pack 5
Temperature Range (Metric) Room Temperature
Dimensions (L x W x H) 128 x 85 x 19 mm
Length (Metric) 127.76 mm
Width (Metric) 85.48 mm
Height (Metric) 19 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.