Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One CELLCOAT™ Poly-L-lysine Coated Cell Culture Dish

Catalog No. 07000586 Shop All Greiner Bio-One Products
Change view
Click to view available options
Diameter (Metric):
60 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
07-000-586 60 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-000-586 Supplier Greiner Bio-One Supplier No. 628930
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For cultivation in serum-free and serum reduced medium. Greiner Bio-One CELLCOAT™ Poly-L-lysine Coated Cell Culture Vessel is used for cultivation of transfected cells and fastidious cell lines, serum-free or -reduced cell culture and general improvement o

  • Source: Synthetic
  • Cultivation of neuronal cells, transfected cells and other fastidious cell lines
  • Cultivation in serum-free and serumreduced medium
  • Improvement of cell attachment
  • Shelf life at room temperature:
TRUSTED_SUSTAINABILITY

Specifications

Closure Color Clear
Closure Included Closure Included
Closure Material Polystyrene
Bottom Shape Flat
Capacity (Metric) 28 mL
Color Clear
Culture Area 21 sq. cm
Material Polystyrene
Shape Round
Sterility Non-sterile
Surface Treatment Poly-L-Lysine
Type Cell Culture Dish
Volume (Metric) Working 6 to 7 mL
Includes Lid
Height (Metric) 15 mm
Diameter (Metric) 60 mm
Show More Show Less

For research use only. Not for use in diagnostic procedures, unless otherwise indicated

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.