Learn More
Description
- Coated with Poly-L-Lysine synthetic protein (30-70 kDa)
- For promotion of cell adhesion, proliferation and growth of transfected cell lines, neuronal cell lines, primary neurons and glial cells
- Ready-to-use products
- Includes lid with condensation rings
Specifications
Specifications
| Capacity (English) | 8.45 oz. |
| Closure Color | Red |
| Closure Material | Polystyrene |
| Color | Clear |
| Capacity (Metric) | 250 mL |
| Material | Polystyrene |
| For Use With (Application) | Low-serum cultivation, serum-free cultivation, cell differentiation, cell adhesion, cell proliferation, growth of transfected cell lines, growth of neuronal cell lines, growth of primary neurons, growth of glial cells |
| Neck Style | Canted |
| Product Type | Cell Culture Flask |
| Surface Coating | Poly-L-Lysine |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.