Learn More
Sigma Aldrich Fine Chemicals Biosciences CellCrown(TM) inserts 6 well plate inserts with 1.0 mum polycarbonate filter, sterile | 3EA
Catalog No. NC1929253
Encompass
CellCrown(TM) inserts 6 well plate inserts with 1.0 mum polycarbonate filter, sterile | 3EA
Catalog No. NC1929253 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z6818223EA
CellCrown(TM) inserts 6 well plate inserts with 1.0 mum polycarbonate filter, sterile | 3EA