Learn More
Sigma Aldrich Fine Chemicals Biosciences CellCrownTM inserts 24 wel
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z74238112EA
Encompass
CellCrown is a disposable insert which immobilizes a sample in its place on a 6 12 24 48 and 96-well plate. fibers foils membranes textiles tissue and block like samples can be fixed with CellCrown in a firm adjusted position in the well. The sample can be left in the well during the change of media by means of a pipette between the wall of the well and the CellCrown. The lid of the well plate can be closed while CellCrown is used. The insert can be removed from the well plate and unloaded after cell culturing without damaging the sample. CellCrown is made of medical grade polycarbonate and the product range includes sterile products ready to use as well as non-sterile products to be sterilized by the user. Sterilization can be by gamma or e-beam radiation autoclaving ethylene oxide gas and plasma sterilization.Sterilization ConditionsTemperature.......121 C / 250 fTime..................15 min.Drying time.........15 min.
Catalog No. 50-290-0573