Learn More
CELLCRUSHER CELLCRUSHER KIT WITH TISSUE PU
Catalog No. NC1824866
Encompass
This ergonomically designed tissue pulverizer from Cellcrusher is used with liquid nitrogen to process varied tissue samples. This product option includes the tissue pulverizer and its accessories, a mallet, handling pliers and recovery spoon.
Catalog No. NC1824866 Supplier CELLCRUSHER Supplier No. CELLCRUSHER KIT
This ergonomically designed tissue pulverizer from Cellcrusher is used with liquid nitrogen to process varied tissue samples. This product option includes the tissue pulverizer and its accessories, a mallet, handling pliers and recovery spoon.