Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ CellCube™ Systems with Corner Clips
GREENER_CHOICE

Catalog No. 10320204 Shop All Corning Products
Change view
Click to view available options
Surface Treatment:
Tissue Culture-treated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Surface Treatment
10-320-204 Tissue Culture-treated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10-320-204 Supplier Corning™ Supplier No. 3264
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The CellCube™ module is an integral, encapsulated, sterile, single-use device that is 100% pressure-hold tested before shipment. It is comprised of a series of parallel, styrene plates joined to create thin, sealed, laminar flow spaces between adjacent plates. Each culture plate receives proprietary tissue culture treatment prior to assembly to ensure dense, uniform cell growth.

CellCube™ modules provide:

  • a series of parallel, polystyrene plates joined to create thin, sealed, laminar flow spaces between adjacent plates.
  • 85,000 cm2 polystyrene growth surface optimally treated with the Corning™ CellBIND™ surface for the growth of attachment dependent cells.
  • a perfused environment which more closely simulates in vivo conditions and reliably distributes nutrients and oxygen.
  • an integral, encapsulated, sterile, single-use device that is 100% pressure-hold tested before shipment.

Corning™ CellBIND™ Surface Benefits:

  • Improves cell attachment leading to increased cell growth and yields
  • Results in more consistent and even cell attachment
  • Helps adapt cells to reduced serum or serum-free conditions
  • Reduces premature cell detachment from confluent cultures
  • May eliminate the need for tedious, time-consuming, expensive and low stability biological coatings
  • Not a coating, requires no refrigeration or special handling and is stable at room temperature
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 6 L
Material Polycarbonate
For Use With (Application) Mass Culture of Attachment-dependent Cells
For Use With (Equipment) CellCube System, Eppendorf BioFlo 120 Controller, and BioBLU 3c Single-use Bioreactor
Includes Corner clips
No. per Case 1/Cs.
Product Type CellCube Module
Shape Square
Disposable Single-use
Graduated No
Sterility Sterile
Culture Area 85,000 cm2
Surface Treatment Tissue Culture-treated
Tray Layers 100
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.