Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ CELLect™ Gold Fetal Bovine Serum, US Origin

Catalog No. MP092916754 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP092916754 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP092916754 Supplier MP Biomedicals™ Supplier No. 092916754
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Superior fetal bovine serum for hybridoma cloning, collected from a U.S.D.A. regulated and monitored abattoir via cardiac puncture. Sterilized via filtration through multiple 0.1 micron filters.

  • An extremely complex mixture of many small and large biomolecules with different, physiologically balanced growth-promoting and growth inhibiting activities
  • CELLect™ Gold is collected from a U.S.D.A. regulated and monitored abattoir by technicians who work on-site (where the fetal blood is aseptically collected via cardiac puncture)
  • Raw material is transported to the production facility where it undergoes sterilization via filtration through multiple 0.1 micron filters
  • The care and diligence taken in handling the fetal serum minimizes cell lysis and other detrimental factors
  • Reduced lysozyme and hemoglobin contamination translates into increased performance in the final product
  • Serum is superior for hybridoma cloning
  • Supplied sterile, with no detectable mycoplasma or viral infection
TRUSTED_SUSTAINABILITY

Specifications

Description FBS Media Supplement
Species Bovine
Country of Origin United States
Form Liquid
Additive Media Supplement - Serum
Product Type Fetal Bovine Serum
Quantity 500 mL
Sterility Sterile
Sterilization Method Sequential-filtered, 100 nm
Tested For Mycoplasma, Viral Infection
Volume (Metric) 500 mL
Formulation Liquid - Glucose: 116 mg/dL, Sodium: 140 mEq/L, BUNN: 13 mg/dL, Chloride: 100 mEq/L, Creatinine: 2.4 mg/dL, Uric Acid: 3.2 mg/dL, Calcium: 11.9 mg/dL, Phosphorus: 8 mg/dL, Albumin: 2.6 g/dL, Total Bilirubin: 0.3 mg/dL, Triglyceride: 32 mg/dL, Alkaline Phosphatase: 168 U/L, AST (SGOT): 35 U/L, LDH: 585 U/L, Cholesterol: < 40 mg/dL, Triglycerides: 0.43 mmol/L, IgG Content: < 60 mg/L, GGT: 7 U/L, Electrophoretic Pattern Normal Alpha1: 3 g/L, Alpha 2: 11 g/L, Beta: 4 g/L, and Gamma: 1 g/L
For Use With (Application) Mammalian Cell Culture and Stem Cell Culture
Lyophilized No
Mycoplasma Detected Not Detected
pH 6.7 to 7.8
Protein Content 3.1 to 4.4 g/dL
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.