Learn More
Bellco Glass, Inc. Cellector Complete,85 mL
SDP
Catalog No. 501945227
Encompass_Preferred
Description: Cellector Complete, 85mL. MADE IN THE USA•Tissue Sieve Homogenizer, used to prepare cell suspensions and gel extractions of proteins•All stainless steel construction•Pan Dimensions: 1-1/4” OD x 2-1/2” W•Complete consists of: Cellector pan & key, glass pestle, 9 screens; 1 Ea from 10 to 150 mesh
Catalog No. 50-194-5227 Supplier Bellco Glass, Inc. Supplier No. 198585000
Description: Cellector Complete, 85mL. MADE IN THE USA•Tissue Sieve Homogenizer, used to prepare cell suspensions and gel extractions of proteins•All stainless steel construction•Pan Dimensions: 1-1/4” OD x 2-1/2” W•Complete consists of: Cellector pan & key, glass pestle, 9 screens; 1 Ea from 10 to 150 mesh