Learn More
Bellco Glass, Inc. Cellector Key Only
SDP
Catalog No. 501945226
Encompass_Preferred
Description: Cellector Key Only. MADE IN THE USA•Cellector key allows the user to secure and detach mesh screens from Cellector Tissue Sieve Homogenizer with ease•All 304 stainless steel construction•Pan Dimensions: 1-1/4” OD x 2-1/2” W•Compatible with 85mL
Catalog No. 50-194-5226 Supplier Bellco Glass, Inc. Supplier No. 198528500
Description: Cellector Key Only. MADE IN THE USA•Cellector key allows the user to secure and detach mesh screens from Cellector Tissue Sieve Homogenizer with ease•All 304 stainless steel construction•Pan Dimensions: 1-1/4” OD x 2-1/2” W•Compatible with 85mL