Learn More
ILEX LIFE SCIENCES LLC Cellendes 3D Life 10x CB Buffer Phenol red free 600 ul 600 UL 1/EA
SDP
Catalog No. 5000580394
Encompass_Preferred
3-D Life 10x CB Buffer (pH 7 2) Phenol Red-Free (600 l) Hydrogel Component DESCRIPTION 3-D Life 10x CB-Buffer (pH 7 2) Phenol Red-Free is a component of the 3-D Life Hydrogel system In order to avoid interferences with light microscopy analysis this buffer does not contain Phenol Red It is a 10x concentrated salt and buffer solution which is diluted to physiological strength when used in the 3-D Life Hydrogel system It is used for the formation of slow gelling 3-D Life Hydrogels where it provides a cell compatible environment at pH 7 2 In conjunction with 3-D Life 10x CB-Buffer (pH 5 5) it is used to generate pHs of intermediate values to modify the gelation kinetics of fast gelling gels APPLICATION Used with other components of the 3-D Life Hydrogel System to establish 3D cell cultures QUANTITY 600 l (sufficient for at least 6 ml 3-D Life Hydrogel)
Catalog No. 50-005-80394 Supplier ILEX LIFE SCIENCES LLC Supplier No. B213
3-D Life 10x CB Buffer (pH 7 2) Phenol Red-Free (600 l) Hydrogel Component DESCRIPTION 3-D Life 10x CB-Buffer (pH 7 2) Phenol Red-Free is a component of the 3-D Life Hydrogel system In order to avoid interferences with light microscopy analysis this buffer does not contain Phenol Red It is a 10x concentrated salt and buffer solution which is diluted to physiological strength when used in the 3-D Life Hydrogel system It is used for the formation of slow gelling 3-D Life Hydrogels where it provides a cell compatible environment at pH 7 2 In conjunction with 3-D Life 10x CB-Buffer (pH 5 5) it is used to generate pHs of intermediate values to modify the gelation kinetics of fast gelling gels APPLICATION Used with other components of the 3-D Life Hydrogel System to establish 3D cell cultures QUANTITY 600 l (sufficient for at least 6 ml 3-D Life Hydrogel)