Learn More
ILEX LIFE SCIENCES LLC Cellendes 3D Life Dextran CD HA Slow Gelling 1/EA
SDP
Catalog No. 5000580396
Encompass_Preferred
Cellendes 3-D Life Dextran-CD-HA Hydrogel Ready-To-Design slow-gelling biomimetic hydrogel kit for 3D cell cultures DESCRIPTION Dextran-CD-HA Hydrogel Kit allows formation of cell-degradable hydrogels containing hyaluronic acid (HA) CD-HyLink HA crosslinker forms stable thioether bonds with thiol-reactive groups on SG-Dextran which results in the formation of the gel The components are mixed at physiological pH (pH 7 2) for optimal cell compatibility The slow gelation kinetics allows enough time to conveniently manipulate the solution before the onset of gel formation CD-HyLink contains an MMP-cleavable peptide sequence which allows cells to locally degrade the gel if they produce the indicated MMPs Cell adhesion peptides (e g 3-D Life RGD Peptide) can be covalently attached to a SG-Dextran to provide a cell-adhesive matrix QUANTITY One kit allows formation of 3 3 ml 3-D Life Hydrogel of a crosslinking strength of 1 2 mmol/L
Catalog No. 50-005-80396 Supplier ILEX LIFE SCIENCES LLC Supplier No. G961
Add to Cart
Add to Cart
Cellendes 3-D Life Dextran-CD-HA Hydrogel Ready-To-Design slow-gelling biomimetic hydrogel kit for 3D cell cultures DESCRIPTION Dextran-CD-HA Hydrogel Kit allows formation of cell-degradable hydrogels containing hyaluronic acid (HA) CD-HyLink HA crosslinker forms stable thioether bonds with thiol-reactive groups on SG-Dextran which results in the formation of the gel The components are mixed at physiological pH (pH 7 2) for optimal cell compatibility The slow gelation kinetics allows enough time to conveniently manipulate the solution before the onset of gel formation CD-HyLink contains an MMP-cleavable peptide sequence which allows cells to locally degrade the gel if they produce the indicated MMPs Cell adhesion peptides (e g 3-D Life RGD Peptide) can be covalently attached to a SG-Dextran to provide a cell-adhesive matrix QUANTITY One kit allows formation of 3 3 ml 3-D Life Hydrogel of a crosslinking strength of 1 2 mmol/L