Learn More
SANISURE CellGyn TPE Tubing 3/8" ID X 5/8" OD
Catalog No. NC3781727
Encompass_Preferred
CellGyn is a high-performance TPE medical tubing engineered specifically for the most demanding biopharma fluid processing environments. As a purpose-built weldable TPE tubing, CellGyn is ideal for single-use assemblies, aseptic transfer lines, sampling devices, and any application requiring reliable aseptic tube welding or sealing. Compatible with all major tube sealers and aseptic tube welders – including Vante, Terumo, BioWelder, and the GE Sterile Tube Fuser – CellGyn delivers superior gas barrier properties and pressure performance that consistently outperforms standard silicone tubing. 
Catalog No. NC3781727 Supplier SANISURE Supplier No. CGN375X62550
CellGyn is a high-performance TPE medical tubing engineered specifically for the most demanding biopharma fluid processing environments. As a purpose-built weldable TPE tubing, CellGyn is ideal for single-use assemblies, aseptic transfer lines, sampling devices, and any application requiring reliable aseptic tube welding or sealing. Compatible with all major tube sealers and aseptic tube welders – including Vante, Terumo, BioWelder, and the GE Sterile Tube Fuser – CellGyn delivers superior gas barrier properties and pressure performance that consistently outperforms standard silicone tubing. 