Learn More
SANISURE CellGyn TPE Tubing 5/16" ID X 7/16" OD
Catalog No. NC2832118
Encompass_Preferred
CellGyn is a high-performance TPE medical tubing engineered specifically for the most demanding biopharma fluid processing environments. As a purpose-built weldable TPE tubing, CellGyn is ideal for single-use assemblies, aseptic transfer lines, sampling devices, and any application requiring reliable aseptic tube welding or sealing. Compatible with all major tube sealers and aseptic tube welders – including Vante, Terumo, BioWelder, and the GE Sterile Tube Fuser – CellGyn delivers superior gas barrier properties and pressure performance that consistently outperforms standard silicone tubing. 
Catalog No. NC2832118 Supplier SANISURE Supplier No. CGN312X43750
CellGyn is a high-performance TPE medical tubing engineered specifically for the most demanding biopharma fluid processing environments. As a purpose-built weldable TPE tubing, CellGyn is ideal for single-use assemblies, aseptic transfer lines, sampling devices, and any application requiring reliable aseptic tube welding or sealing. Compatible with all major tube sealers and aseptic tube welders – including Vante, Terumo, BioWelder, and the GE Sterile Tube Fuser – CellGyn delivers superior gas barrier properties and pressure performance that consistently outperforms standard silicone tubing. 