Learn More
StatLab CellNas SmartStor Supa Mega Block Storage Box
SDP
Catalog No. 22000091
Change view
Click to view available options
Color:
Light Brown
Catalog No. Color
22-000-091 Light Brown
1 options
Catalog No. 22-000-091 Supplier StatLab Supplier No. WSD040008A
Holds up to 60 Supa Mega Cassettes
  • Manufactured from recycled fiberboard, the SmartStor Supa Mega Block Archive Box is an economical solution to the long-term archiving of Supa Mega blocks
  • Cost-effective archiving solution
  • Accommodates 60 Supa Mega, 128 Supa Mega Slim or 80 Supa Mega Mothership blocks
  • Compatible with the CellNass SmartStor 2 Cabinet
Specifications
Product Type Cardboard Block Filing Cabinet
Color Light Brown
Holds 60 to 128 Mega Cassettes