Learn More
Apexbio Technology LLC Cellobiose 528-50-7 100mg
SDP
Catalog No. 502240468
Encompass_Preferred
Cellobiose (CAS 528-50-7) is a disaccharide composed of two -D-glucose units linked by a (1 4) glycosidic bond It functions as a reducing sugar and is cleaved into glucose by -glucosidase enzymes or acidic hydrolysis Research indicates that oral administration of cellobiose can attenuate clinical and histopathological indicators of colitis in animal models accompanied by decreased mucosal expression of pro-inflammatory cytokine mRNAs These findings support its utility in studies of intestinal inflammation digestion and carbohydrate metabolism
This item is not returnable. View return policy
Catalog No. 50-224-0468 Supplier Apexbio Technology LLC Supplier No. B7879100
Cellobiose (CAS 528-50-7) is a disaccharide composed of two -D-glucose units linked by a (1 4) glycosidic bond It functions as a reducing sugar and is cleaved into glucose by -glucosidase enzymes or acidic hydrolysis Research indicates that oral administration of cellobiose can attenuate clinical and histopathological indicators of colitis in animal models accompanied by decreased mucosal expression of pro-inflammatory cytokine mRNAs These findings support its utility in studies of intestinal inflammation digestion and carbohydrate metabolism