Learn More
World Precision Instrument Celloger Pro
SDP
Catalog No. NC2819334
Encompass
Celloger Pro is an innovative live cell imaging system that redefines research capabilities. With its exceptional image quality and unmatched convenience, it empowers researchers with advanced features. By enabling real-time cell monitoring inside the incubator, it allows for seamless observation and tracking of cellular dynamics without disrupting the natural growth environment. The system's dual fluorescence and bright-field microscopy enable simultaneous visualization of multiple markers, while the multi-point time-lapse imaging captures dynamic cellular events across different locations.     NOTE: WPI is an authorized distributor of 
Catalog No. NC2819334 Supplier World Precision Instrument Supplier No. EVILCI0108
Add to Cart
Add to Cart
Request a Quote
Celloger Pro is an innovative live cell imaging system that redefines research capabilities. With its exceptional image quality and unmatched convenience, it empowers researchers with advanced features. By enabling real-time cell monitoring inside the incubator, it allows for seamless observation and tracking of cellular dynamics without disrupting the natural growth environment. The system's dual fluorescence and bright-field microscopy enable simultaneous visualization of multiple markers, while the multi-point time-lapse imaging captures dynamic cellular events across different locations.     NOTE: WPI is an authorized distributor of 