Learn More
Electron Microscopy Sciences Cellometer™ Disposable Counting Chamber, Plastic Base
SDP
Catalog No. 5031118
Encompass_Preferred
These Disposable Cell Counters are an enclosed chamber with two ports for sample introduction. The chamber contains precisely spaced lines in a grid pattern. There are two counting chambers per slide for replicates. The counting grid pattern is the Improved Neubauer, as in a common Hemacytometer. The consistent pattern design allows the standard cell counting procedure to be followed.
Catalog No. 50-311-18 Supplier Electron Microscopy Sciences Supplier No. 6356125
These Disposable Cell Counters are an enclosed chamber with two ports for sample introduction. The chamber contains precisely spaced lines in a grid pattern. There are two counting chambers per slide for replicates. The counting grid pattern is the Improved Neubauer, as in a common Hemacytometer. The consistent pattern design allows the standard cell counting procedure to be followed.