Learn More
Bellco Glass, Inc. Cellometer Plastic,1/2x3-Dual Dual-Pack 25 per cs
SDP
Supplier:  Bellco Glass, Inc. 563820125
Encompass_Preferred
Cellometer plastic disposable cell counting chamber features an improved neubauer counting grid on plastic that is mounted onto a standard 25x75mm plastic slide. The dual counting chamber has a sample introduction port, counting grid and air evacuation port & is ideal for laboratories working with infectious samples. duplicate samples may be counted, or control/unknown samples may be counted on the same slide. no coverslip is required for the cellometer thus avoiding counting errors. pack 25/per cs
Catalog No. 50-194-5830