Learn More
Electron Microscopy Sciences 14 x 14 cm Cellophane, 50 sheets
SDP
Catalog No. 5034527
Encompass_Preferred
Cellophane for drying frames, 50 pack. 14 x 14 cm. Gel Drying Frames allow researchers to quickly and easily dry gels between two sheets of cellophane without the use of expensive heat/vacuum gel dryers. Use with the Gel Drying Frames. *Cellophane only*
Catalog No. 50-345-27 Supplier Electron Microscopy Sciences Supplier No. 6851114
Cellophane for drying frames, 50 pack. 14 x 14 cm. Gel Drying Frames allow researchers to quickly and easily dry gels between two sheets of cellophane without the use of expensive heat/vacuum gel dryers. Use with the Gel Drying Frames. *Cellophane only*