Learn More
Cellosolve™ 2-Ethoxyethanol, ≥99.8% (GC), MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsDescription
Specifications
Specifications
| Chemical Name or Material | 2-Ethoxyethanol |
| CAS | 110-80-5 |
| Product Line | Cellosolve |
| Ignition Point | 460°F |
| Vapor Pressure | 3.8 mmHg (20°C) |
| Vapor Density | 3.1 (vs. air) |
| Density | 0.93 g/mL (at 25°C) |
| Flash Point | 42°C |
| Physical Form | Neat |
| Linear Formula | C2H5 OCH2 CH2 OH |
| Show More |
Cellosolve is a registered trademark of Union Carbide
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.