Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cellosolve™ 2-Ethoxyethanol Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111017849 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
3 x 1.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-7849 3 x 1.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-7849 Supplier MilliporeSigma Supelco Supplier No. PHR15503X1.2ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
Product Line Cellosolve
Ignition Point 460°F
CAS 110-80-5
Vapor Pressure 3.8 mmHg (20°C)
Vapor Density 3.1 (vs. air)
Density 0.93 g/mL (at 25°C)
Flash Point 42°C - Closed Cup
Linear Formula C2H5 OCH2 CH2 OH
MDL Number MFCD00002869
Melting Point -90°C
Boiling Point 135°C
Synonym 2-Ethoxyethanol; Cellosolve(R); Ethyl glycol; Ethylene glycol monoethyl ether
Type Certified Reference Material
Quantity 3 x 1.2 mL
Formula Weight 90.12
Grade Certified Reference Material
Show More Show Less

Cellosolve is a registered trademark of Union Carbide

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.