Learn More
Iwai North America Cellotion: Serum, protein, and sugar free chemically defined cell washing solution. Size: 100mL
Catalog No. NC2733012
Encompass_Preferred
Cellotion is a chemically defined cell washing solution that significantly increase cell yield after washing procedures while maintaining cell viability. Its unique features, free of serum, protein, and sugars, is ideal for cells, tissues, and organs.
Catalog No. NC2733012 Supplier Iwai North America Supplier No. 11918
Cellotion is a chemically defined cell washing solution that significantly increase cell yield after washing procedures while maintaining cell viability. Its unique features, free of serum, protein, and sugars, is ideal for cells, tissues, and organs.