Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cellotriose 95.0+%, TCI America™
SDP

Catalog No. C279520MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
20 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C279520MG 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C279520MG Supplier TCI America Supplier No. C279520MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 33404-34-1
Molecular Formula C18H32O16
Molecular Weight (g/mol) 504.44
MDL Number MFCD00069842
InChI Key FYGDTMLNYKFZSV-ZWSAEMDYSA-N
Synonym cellotriose, d-+-cellotriose, polysaccharide, d-cellotriose, beta-d-glucopyranosyl-1->4-beta-d-glucopyranosyl-1->4-d-glucose, beta-d-glcp-1->4-beta-d-glcp-1->4-d-glc, beta-d-glucopyranosyl-1->4-beta-d-glucopyranosyl-1->4-d-glucopyranose, cellodextrin, d-glucose, o-d-glucopyranosyl, cellooligosaccharide
PubChem CID 5287993
ChEBI CHEBI:3528
IUPAC Name (2S,3R,4S,5S,6R)-2-{[(2R,3S,4R,5R,6S)-4,5-dihydroxy-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxan-3-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](O[C@H]3[C@H](O)[C@@H](O)C(O)O[C@@H]3CO)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Melting Point 209°C
Color White
Quantity 20 mg
Formula Weight 504.44
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Cellotriose
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.