Learn More
Cellpath Acid Resistant Polyester Biopsy Bags
SDP
Catalog No. 22222013
Change view
Click to view available options
Dimensions (L x W):
40 x 30 mm
Catalog No. Dimensions (L x W)
22-222-013 40 x 30 mm
1 options
This item is not returnable. View return policy
Catalog No. 22-222-013 Supplier Cellpath Supplier No. EBF010002P
This item is not returnable. View return policy
Holds biopsy sample during processing. CellPath Acid Resistant Polyester Biopsy Bags are great alternatives to BioWrap paper and foam biopsy pads. Particularly helpful when collecting endometrial samples from formalin as it can be used as a sieve.
  • Easy tear for quick sample retrieval
  • Acid resistant polyester useful when processing very fine bone samples
  • Useful for collecting endometrial samples from formalin
  • Time saving
Specifications
Material Polyester
Dimensions (L x W) 40 x 30 mm
Quantity 100/Pk.