Learn More
Cellpath BioPad Biopsy Foam Pads
Catalog No. 22222012
Change view
Click to view available options
Color:
Blue
Catalog No. Color
22-222-012 Blue
1 options
This item is not returnable. View return policy
Catalog No. 22-222-012 Supplier Cellpath Supplier No. EBA010603A
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Allow fluid to thoroughly and safely transfer around the tissue sample, whilst also holding the tissue specimen safely in place. CellPath Biopsy Foam Pads can be cut to size to fit most major processing cassettes.
  • Cut to size to fit most major processing cassettes
  • Prevents loss of small specimens
  • Low cost
  • Efficient
Specifications
Color Blue