Learn More
Cellpath CellSafe+ Blue Biopsy Capsules
SDP
Catalog No. 22222007
Change view
Click to view available options
Quantity:
100/Pk.
1000/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-222-007 100/Pk.
22-222-008 1000/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-222-007 Supplier Cellpath Supplier No. EBE030102A
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Offers high security and easy to use alternative to traditional biopsy handling techniques such as tissue wrap and foam pads which are proving to be unsatisfactory and time consuming. CellPath CellSafe+ Blue Biopsy Capsules features new blue mesh that provides better contrast between the biopsy and the CellSafe.
  • Features two hinged interlocking frames with an integral nylon mesh
  • Adjustable to biopsy specimen size
  • Hinged in either direction and closed to encapsulate the tissue
  • New Blue Mesh provides better contrast between the biopsy and the CellSafe
  • X–Ray transparent mesh for breast work
  • Extra fine mesh provides up to 20 times less carry–over of solvents than foam pads
  • Zero Tissue damage or artefacts associated with foam pads
  • Substantial time savings compared to folding biopsy in tissue paper
Specifications
Quantity 100/Pk.
Color Blue