Learn More
StatLab CellPath Mini Slide Dryer Base
SDP
Catalog No. 23640026
Change view
Click to view available options
Slide Capacity:
150
Catalog No. Slide Capacity
23-640-026 150
1 options
Catalog No. 23-640-026 Supplier StatLab Supplier No. JAX030100A
Add to Cart
Add to Cart
Designed for microtome use, delivering rapid, efficient section drying with digitally controlled airflow.
  • Purpose-designed Mini Slide Dryer Base for use at the microtome, supporting improved LEAN workflow by enabling immediate and efficient slide drying directly at the point of sectioning.
  • Digitally controlled airflow system delivers warm air evenly through the base of the instrument, ensuring rapid and consistent section drying while minimising the risk of specimen overheating or damage.
  • Efficient drying process significantly reduces overall drying times by approximately 75% when compared with conventional laboratory ovens, improving turnaround time and productivity.
Specifications
Product Type Microscope Slide Dryer
Slide Capacity 150