Learn More
StatLab CellPath Mini Slide Dryer Rack
SDP
Catalog No. 23640028
Change view
Click to view available options
Slide Capacity:
20 to 30/Tray
Catalog No. Slide Capacity
23-640-028 20 to 30/Tray
1 options
Catalog No. 23-640-028 Supplier StatLab Supplier No. JAX030000C
Add to Cart
Add to Cart
Optional accessory for the CellPath Mini Slide Dryer, designed to support accurate, timed drying when used with the LEAN Timer System.
  • Compatible slide rack accessory for the CellPath Mini Slide Dryer, designed for use with the optional LEAN Timer System and identifier rack to ensure each slide rack is assigned an exact, repeatable drying time for consistent laboratory slide drying results.
  • Works in conjunction with the LEAN Timer System to provide clear audible and visual notifications when a slide rack has fully completed the drying cycle, helping users reduce errors, improve workflow control, and avoid incomplete or excessive drying.
Specifications
Product Type Slide Warmer Tray
Slide Capacity 20 to 30/Tray