Learn More
StatLab CellPath Mini Slide Dryer Timer
SDP
Catalog No. 23640027
Change view
Click to view available options
Slide Capacity:
150
Catalog No. Slide Capacity
23-640-027 150
1 options
Catalog No. 23-640-027 Supplier StatLab Supplier No. JAX030000B
Add to Cart
Add to Cart
Optional LEAN timer accessory for the CellPath Mini Slide Dryer, providing precise slide rack drying control with clear completion alerts
  • LEAN Timer System accessory designed for the CellPath Mini Slide Dryer to ensure each slide rack is assigned and monitored with an exact, repeatable drying time, improving workflow efficiency and consistency in laboratory slide drying.
  • Integrated identifier rack supports accurate rack tracking, while combined audible and visual alerts clearly notify users when the drying cycle is fully complete, reducing errors and unattended over-drying.
Specifications
Product Type Dryer Timer
Slide Capacity 150