Learn More
Cellpath ProMarc Supa Mega Mother Ship Tissue Processing Cassettes
SDP
Catalog No. 22222126
Change view
Click to view available options
Color:
Blue
Green
Pink
White
Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-222-126 Yellow
22-222-122 White
22-222-123 Green
22-222-124 Blue
22-222-125 Pink
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 22-222-126 Supplier Cellpath Supplier No. EAO010902A
Add to Cart
Add to Cart
This item is not returnable. View return policy
Provides an innovative solution for the processing and embedding of large tissue specimens. CellPath ProMarc Supa Mega Mother Ship Tissue Processing Cassettes feature an internal chamber a third of the depth of the conventional Supa Mega Cassette.
  • Allows use of standard tissue processing cassette to clearly print 2D barcode and test infomation
  • Designed using a docking area for standard processing cassette
  • Strong Hex pore structure improves processing for large specimens
  • Reduces turn around times
  • Time saving by ensuring rapid casting of block, 30 minutes at -8°C
  • Thinner chamber depths reduce specimen thickness and improves specimen quality by reducing distortion with no curling
  • Better sectioning quality through adhesion of paraffin block to cassette
  • Available in various colors
Specifications
Quantity 500/Pk.
Color Yellow