Learn More
Alkali Scientific CellPro™ 0.36 ml 96-Well Deep Well Plate, V-Bottom, Round well, 5/pk, 50/cs, Sterile
SDP
Catalog No. 503029444
Encompass_Preferred
Our 0.36 ml 96-Well Deep Well Plate features a V-Bottom design with round wells. Each pack contains 5 sterile plates, and there are 50 packs in a case. This product is ideal for various laboratory applications requiring cell culture, sample storage, or assay screening. The V-Bottom design allows for easy sample concentration and reconstitution, making it efficient for multiple research tasks. The sterile feature ensures aseptic conditions for cell cultures and minimizes potential contamination risks. With a total capacity of 0.36 ml, these plates are suitable for handling small sample volumes, promoting cost-effectiveness by reducing reagent waste. Whether used for liquid handling procedures or storage purposes, these deep well plates offer reliability and convenience in a lab setting. 
Catalog No. 50-302-9444 Supplier Alkali Scientific Supplier No. MPD30S
Our 0.36 ml 96-Well Deep Well Plate features a V-Bottom design with round wells. Each pack contains 5 sterile plates, and there are 50 packs in a case. This product is ideal for various laboratory applications requiring cell culture, sample storage, or assay screening. The V-Bottom design allows for easy sample concentration and reconstitution, making it efficient for multiple research tasks. The sterile feature ensures aseptic conditions for cell cultures and minimizes potential contamination risks. With a total capacity of 0.36 ml, these plates are suitable for handling small sample volumes, promoting cost-effectiveness by reducing reagent waste. Whether used for liquid handling procedures or storage purposes, these deep well plates offer reliability and convenience in a lab setting. 