Learn More
Alkali Scientific CellPro™ 0.5mL 96-Well, Deep Well Plate, V Conical Bottom, Square Well, for KingFisher™ Extraction Platform, 50/case, DNase, RNase & Pyrogen-Free
SDP
Catalog No. 503026746
Encompass_Preferred
Low binding medical grade polypropylene construction. For use with Viral RNA extraction, DNA extraction, RNA/DNA quantification, and immunopreciptitations.
Catalog No. 50-302-6746 Supplier Alkali Scientific Supplier No. MPD22
Add to Cart
Add to Cart
Low binding medical grade polypropylene construction. For use with Viral RNA extraction, DNA extraction, RNA/DNA quantification, and immunopreciptitations.