Learn More
Alkali Scientific CellPro 0.5mL Microcentrifuge
SDP
Catalog No. 503027233
Encompass_Preferred
The Amber 0 5mL Microcentrifuge Tubes are specifically designed for secure handling and storage of small-volume light-sensitive samples Constructed from durable amber polypropylene these tubes effectively block light to protect sensitive reagents and samples The conical design ensures efficient sample retrieval while the innovative lock cap provides a secure seal for added reliability during high-speed centrifugation and storage Amber Polypropylene Reduces light penetration to preserve the integrity of photosensitive samples and reagents Lock Cap Design Provides a leak-proof seal while remaining easy to open and close Compact Size Ideal for small-volume applications in molecular biology biochemistry and related fields High Centrifugal Strength Engineered to withstand rigorous centrifugation conditions Frosted Marking Area Conveniently labeled tubes for easy identification and organization Versatile Use Perfect for light-sensitive sample storage chemiluminescence reagents and ot
Catalog No. 50-302-7233 Supplier Alkali Scientific Supplier No. CN3006AMB
The Amber 0 5mL Microcentrifuge Tubes are specifically designed for secure handling and storage of small-volume light-sensitive samples Constructed from durable amber polypropylene these tubes effectively block light to protect sensitive reagents and samples The conical design ensures efficient sample retrieval while the innovative lock cap provides a secure seal for added reliability during high-speed centrifugation and storage Amber Polypropylene Reduces light penetration to preserve the integrity of photosensitive samples and reagents Lock Cap Design Provides a leak-proof seal while remaining easy to open and close Compact Size Ideal for small-volume applications in molecular biology biochemistry and related fields High Centrifugal Strength Engineered to withstand rigorous centrifugation conditions Frosted Marking Area Conveniently labeled tubes for easy identification and organization Versatile Use Perfect for light-sensitive sample storage chemiluminescence reagents and ot