Learn More
Alkali Scientific CellPro™ 1.0 mL 96 Well Deep Well Plate, V Bottom, Round Well, 5/pk, 50/cs, Sterile
SDP
Catalog No. 5000662267
Encompass_Preferred
CellPro™ 1.0 mL 96 well deep well plate designed for reliable sample preparation, storage, and processing in laboratory workflows, with durable polypropylene construction, with V bottom round wells that allow efficient sample recovery and mixing, with a maximum working volume of 1.0 mL per well suitable for biological and chemical applications, with uniform well geometry to support consistent pipetting and automated handling systems, with sterile manufacturing to reduce contamination risk during sensitive procedures, with compatibility for centrifugation, storage, and reagent preparation tasks, with strong structural stability that resists deformation during routine laboratory use, and with packaging of 5 plates per pack and 50 plates per case for convenient laboratory supply management.
Catalog No. 50-006-62267 Supplier Alkali Scientific Supplier No. MPD20S
CellPro™ 1.0 mL 96 well deep well plate designed for reliable sample preparation, storage, and processing in laboratory workflows, with durable polypropylene construction, with V bottom round wells that allow efficient sample recovery and mixing, with a maximum working volume of 1.0 mL per well suitable for biological and chemical applications, with uniform well geometry to support consistent pipetting and automated handling systems, with sterile manufacturing to reduce contamination risk during sensitive procedures, with compatibility for centrifugation, storage, and reagent preparation tasks, with strong structural stability that resists deformation during routine laboratory use, and with packaging of 5 plates per pack and 50 plates per case for convenient laboratory supply management.