Learn More
Alkali Scientific CellPro 1.3mL 96-Well Deep Wel
SDP
Catalog No. 503028686
Encompass_Preferred
1 3mL 96 Well Round Deep Well Plates with ISO9001and13485 certified 1 3ml 96 Well Round Deep Well Plates are manufactured in 100000 grade cleaning room with USP 6 grade raw materials The length and width of the deep well plate comply with the SBS international standards The depth of wells is increased based on the appearance and size of normal plates (mainly 96-well and 384-well plates) to enlarge the volume of each well In addition the manufacturing material (currently mostly used is polypropylene (PP)) is changed and the surface treatment process has been improved for this series of deep well plates to adapt to its specific use range 1 Sample storage it can replace the conventional 1 5mL Centrifuge Tube to store samples and can be arranged tidily to save space with large storage capacity and can withstand the -80 refrigerator condition Therefore its also called storage plate 2 Sample processing it can be used in combined with multipass Pipettes high-throughput automatic liq
Catalog No. 50-302-8686 Supplier Alkali Scientific Supplier No. MPD38S
1 3mL 96 Well Round Deep Well Plates with ISO9001and13485 certified 1 3ml 96 Well Round Deep Well Plates are manufactured in 100000 grade cleaning room with USP 6 grade raw materials The length and width of the deep well plate comply with the SBS international standards The depth of wells is increased based on the appearance and size of normal plates (mainly 96-well and 384-well plates) to enlarge the volume of each well In addition the manufacturing material (currently mostly used is polypropylene (PP)) is changed and the surface treatment process has been improved for this series of deep well plates to adapt to its specific use range 1 Sample storage it can replace the conventional 1 5mL Centrifuge Tube to store samples and can be arranged tidily to save space with large storage capacity and can withstand the -80 refrigerator condition Therefore its also called storage plate 2 Sample processing it can be used in combined with multipass Pipettes high-throughput automatic liq