Learn More
Alkali Scientific CellPro™ 1.5mL Screw Tube, conical base, ribbed, Clear, 500/pk, 5000/case
SDP
Catalog No. 503026608
Encompass_Preferred
Volume: 1.5 mLGraduations: Non-GraduatedRibbing (Knurls): RibbedBase: ConicalColor: -00 ClearPackaging: 500/unit, 10 units/case
Catalog No. 50-302-6608 Supplier Alkali Scientific Supplier No. C3024
Volume: 1.5 mLGraduations: Non-GraduatedRibbing (Knurls): RibbedBase: ConicalColor: -00 ClearPackaging: 500/unit, 10 units/case