Learn More
Alkali Scientific CellPro™ 100 mM Sodium Pyruvate solution, 100ml
SDP
Catalog No. 503029110
Encompass_Preferred
An easily accessible carbohydrate source for cell culture media. Part of a high-performance line of nutritional and growth supplements
Catalog No. 50-302-9110 Supplier Alkali Scientific Supplier No. NAPY01
An easily accessible carbohydrate source for cell culture media. Part of a high-performance line of nutritional and growth supplements