Learn More
Alkali Scientific CellPro™ 100 x 1.5mL Tube Storage Box with Hinged Lid, 5/Pk, Orange
SDP
Catalog No. 503027727
Encompass_Preferred
Suitable for refrigerator and freezer storage as low as -80˚C, Tube Storage Box have translucent lids and numbered cavities.  • Polypropylene, withstand -80˚C• 6 types - for 0.2ml to 50ml tubes• Compact & space efficient• Autoclavable• Numbered tube wells• 5 per Pack
Catalog No. 50-302-7727 Supplier Alkali Scientific Supplier No. FS100O
Suitable for refrigerator and freezer storage as low as -80˚C, Tube Storage Box have translucent lids and numbered cavities.  • Polypropylene, withstand -80˚C• 6 types - for 0.2ml to 50ml tubes• Compact & space efficient• Autoclavable• Numbered tube wells• 5 per Pack