Learn More
Alkali Scientific CellPro™ 100X Vitamin solution, 100ml
SDP
Catalog No. 503028487
Encompass_Preferred
Growth supplement for cell culture medium. Supplement serum- and protein-free media with CellPro™ 100X Vitamin Solution.
Catalog No. 50-302-8487 Supplier Alkali Scientific Supplier No. MEM505
Growth supplement for cell culture medium. Supplement serum- and protein-free media with CellPro™ 100X Vitamin Solution.