Learn More
Alkali Scientific CellPro™ 100mm Cell Culture Dish, TC, Sterile 20/pk, Tall Stack, 300/cs
SDP
Catalog No. 502335953
Encompass_Preferred
CellPro™ Cell Culture Dishes are ideal for all types of culture methods where microscopic observation is required. Vacuum Plasma treatment of the growth surface provides an excellent substrate for adherence of a wide range of cell types. Numeric marks on the bottom of each dish provide a guide that allows users to identify the location of cells. They provide a flat transparent surface for distortion-free observation. They are stackable for easy storage and handling, sterilized by E-Beam and are non-pyrogenic.
Catalog No. 50-233-5953 Supplier Alkali Scientific Supplier No. TDN0100
CellPro™ Cell Culture Dishes are ideal for all types of culture methods where microscopic observation is required. Vacuum Plasma treatment of the growth surface provides an excellent substrate for adherence of a wide range of cell types. Numeric marks on the bottom of each dish provide a guide that allows users to identify the location of cells. They provide a flat transparent surface for distortion-free observation. They are stackable for easy storage and handling, sterilized by E-Beam and are non-pyrogenic.