Learn More
Alkali Scientific CellPro™ 10mL Plain Top Centrifuge Tube, Non Graduated, Conical Bottom (V), Non Skirted, External Rim, Bulk Bagged, Polystyrene (PS), 2500/Case
SDP
Catalog No. 5000662682
Encompass_Preferred
The CellPro™ 10 mL Plain Top Centrifuge Tube features a smooth, non graduated surface designed for laboratory workflows where external labeling or custom measurement markings are preferred. Manufactured from clear polystyrene, the tube includes a conical V bottom that promotes efficient pellet formation and complete sample recovery after centrifugation. The plain open top with an external rim improves handling and enables secure sealing with compatible caps or sealing films. Its non skirted base ensures compatibility with standard centrifuge rotors used in many laboratory centrifugation systems. These tubes are DNase and RNase free and non pyrogenic, making them suitable for sensitive biological and molecular biology samples. Bulk packaging supports high throughput laboratory environments and routine sample preparation workflows.
Catalog No. 50-006-62682 Supplier Alkali Scientific Supplier No. CUT010PSCS
The CellPro™ 10 mL Plain Top Centrifuge Tube features a smooth, non graduated surface designed for laboratory workflows where external labeling or custom measurement markings are preferred. Manufactured from clear polystyrene, the tube includes a conical V bottom that promotes efficient pellet formation and complete sample recovery after centrifugation. The plain open top with an external rim improves handling and enables secure sealing with compatible caps or sealing films. Its non skirted base ensures compatibility with standard centrifuge rotors used in many laboratory centrifugation systems. These tubes are DNase and RNase free and non pyrogenic, making them suitable for sensitive biological and molecular biology samples. Bulk packaging supports high throughput laboratory environments and routine sample preparation workflows.