Learn More
Alkali Scientific CellPro™ 10mL Sterile Transport / Mailing Tube, 10 bags 100 tubes, 1000/cs, Screw-Cap, AMBER, 1000/cs
SDP
Catalog No. 503026460
Encompass_Preferred
CellPro™ 10mL Transport tubes, sometimes called mailing tubes, are used in multiple sectors of the laboratory market, including medical, hospital, environmental, independent testing, food testing, and biological research labs. Also suitable for pharmaceutical packaging and applications.   Used to store, transport or freeze samples, are 10mL (16 x 80mm) and are fitted with leak-proof screw-caps. They are made of high-grade polypropylene, and include white printed graduations and marking area. Tubes are self-standing and come packaged in 10 bags 100 tubes Leak-proof, 1.25 turn screw-cap Meet 95kPa transportation guidelines Vivid, printed graduations Screw-caps attached Sterile, RNase, DNase, Pyrogen free  
Catalog No. 50-302-6460 Supplier Alkali Scientific Supplier No. C3582AM
CellPro™ 10mL Transport tubes, sometimes called mailing tubes, are used in multiple sectors of the laboratory market, including medical, hospital, environmental, independent testing, food testing, and biological research labs. Also suitable for pharmaceutical packaging and applications.   Used to store, transport or freeze samples, are 10mL (16 x 80mm) and are fitted with leak-proof screw-caps. They are made of high-grade polypropylene, and include white printed graduations and marking area. Tubes are self-standing and come packaged in 10 bags 100 tubes Leak-proof, 1.25 turn screw-cap Meet 95kPa transportation guidelines Vivid, printed graduations Screw-caps attached Sterile, RNase, DNase, Pyrogen free  