Learn More
Alkali Scientific CellPro™ 1536 Well Black Square ELISA Plate, 10/Pk, 40/Case, Middle Bind/DNase, RNase & Pyrogen Free
SDP
Catalog No. 5000662002
Encompass_Preferred
CellPro™ 1536 Well Black Square ELISA Plate, 10/Pk, 40/Case, Middle Bind/DNase, RNase & Pyrogen Free. Specially designed for critical ELISA applications, this black plate provides a stable surface with middle bind coating. It ensures accurate and reproducible results while being free of DNase, RNase, and pyrogens. This plate is ideal for research where contamination is a concern. The clear cover facilitates easy handling and secure storage.
Catalog No. 50-006-62002 Supplier Alkali Scientific Supplier No. DWP1536BNH
CellPro™ 1536 Well Black Square ELISA Plate, 10/Pk, 40/Case, Middle Bind/DNase, RNase & Pyrogen Free. Specially designed for critical ELISA applications, this black plate provides a stable surface with middle bind coating. It ensures accurate and reproducible results while being free of DNase, RNase, and pyrogens. This plate is ideal for research where contamination is a concern. The clear cover facilitates easy handling and secure storage.