Learn More
Alkali Scientific CellPro™ 1536 Well Clear Square ELISA Plate, 10/Pk, 40/Case, High Bind/ Sterile/with Clear Cover
SDP
Catalog No. 5000662003
Encompass_Preferred
CellPro™ 1536 Well Clear Square ELISA Plate, 10/Pk, 40/Case, High Bind/Sterile with Clear Cover. This clear plate is designed for applications requiring high binding capacity, featuring a transparent surface for precise well detection. Each plate is sterilized, ensuring it is safe for sensitive assays. The high-bind coating guarantees optimal results in research and clinical labs. The clear cover adds convenience for storage and identification.
Catalog No. 50-006-62003 Supplier Alkali Scientific Supplier No. DWP1536CHS
CellPro™ 1536 Well Clear Square ELISA Plate, 10/Pk, 40/Case, High Bind/Sterile with Clear Cover. This clear plate is designed for applications requiring high binding capacity, featuring a transparent surface for precise well detection. Each plate is sterilized, ensuring it is safe for sensitive assays. The high-bind coating guarantees optimal results in research and clinical labs. The clear cover adds convenience for storage and identification.