Learn More
Alkali Scientific CellPro™ 1536 Well Clear Square ELISA Plate, 10/Pk, 40/Case, Middle Bind/DNase, RNase & Pyrogen Free
SDP
Catalog No. 5000662405
Encompass_Preferred
CellPro™ 1536 Well Clear Square ELISA Plate, 10/Pk, 40/Case, Middle Bind/DNase, RNase & Pyrogen Free. Featuring a middle bind coating, this clear square ELISA plate ensures reliable performance in sensitive assays. It is free from DNase, RNase, and pyrogens, making it ideal for contamination-free testing. With 1536 wells, it provides ample space for high-throughput applications. A clear cover enhances ease of use and identification.
Catalog No. 50-006-62405 Supplier Alkali Scientific Supplier No. DWP1536BCNH
CellPro™ 1536 Well Clear Square ELISA Plate, 10/Pk, 40/Case, Middle Bind/DNase, RNase & Pyrogen Free. Featuring a middle bind coating, this clear square ELISA plate ensures reliable performance in sensitive assays. It is free from DNase, RNase, and pyrogens, making it ideal for contamination-free testing. With 1536 wells, it provides ample space for high-throughput applications. A clear cover enhances ease of use and identification.